Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039320.1 | NP_001034409.1 | 2 - 3 | 239 | 99 | |
GenBank mRNA | AJ719316.1 | - | 2 - 3 | 239 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |