Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039453.1 | NP_001034542.1 | 2 - 3 | 550 | 79 | |
GenBank mRNA | AM183252.1 | - | 2 - 3 | 550 | 79 | |
BX931084.1 | - | 2 - 3 | 616 | 79 | ||
BX933585.1 | - | 2 - 3 | 614 | 79 | ||
BX934345.1 | - | 2 - 3 | 615 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |