Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNASET2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039491.1 | NP_001034580.1 | 6 - 7 | 421 | 77 | |
XM_015284062.2 | XP_015139548.1 | 6 - 7 | 3411 | 77 | ||
XM_015284063.2 | XP_015139549.1 | 6 - 7 | 3477 | 77 | ||
XM_015284064.2 | XP_015139550.1 | 6 - 7 | 3414 | 77 | ||
XM_015284065.2 | XP_015139551.1 | 6 - 7 | 430 | 77 | ||
XM_015284066.2 | XP_015139552.1 | 6 - 7 | 3337 | 77 | ||
GenBank mRNA | AJ447620.1 | - | 6 - 7 | 422 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |