Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARRES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204534.4 | NP_989865.1 | 2 - 3 | 316 | 101 | |
GenBank mRNA | AJ307060.2 | - | 2 - 3 | 316 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |