Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204546.2 | NP_989877.2 | 5 - 6 | 660 | 75 | |
XM_015299306.2 | XP_015154792.1 | 5 - 6 | 659 | 75 | ||
GenBank mRNA | AB080727.1 | - | 5 - 6 | 660 | 75 | |
BU355067.1 | - | 1 - 2 | 63 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |