Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205335.3 | NP_990666.1 | 1 - 2 | 152 | 102 | |
GenBank mRNA | BU119521.1 | - | 1 - 2 | 94 | 102 | |
BU269372.1 | - | 1 - 2 | 152 | 102 | ||
BU339400.1 | - | 1 - 2 | 94 | 102 | ||
BU352734.1 | - | 1 - 2 | 86 | 102 | ||
DN851840.1 | - | 1 - 2 | 130 | 102 | ||
X60471.1 | - | 94 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |