Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001311.2 | NP_001001311.2 | 2 - 3 | 628 | 67 | |
GenBank mRNA | AY574987.1 | - | 2 - 3 | 628 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |