Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001472.2 | NP_001001472.2 | 3 - 4 | 631 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |