Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007977.3 | NP_001007978.1 | 7 - 8 | 928 | 67 | |
GenBank mRNA | CR523513.1 | - | 7 - 8 | 857 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |