Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Gg03359334_g1
See other WDSUB1 GE Assays ›
Gene Symbol:
WDSUB1
Gene Name:
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 36520588 - 36550929 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID
Gg03359334_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: WDSUB1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001031263.2
NP_001026434.2
7 - 8
1175
104
XM_015290154.1
XP_015145640.1
6 - 7
1247
104
XM_015290155.1
XP_015145641.1
6 - 7
1247
104
XM_015290156.1
XP_015145642.1
6 - 7
1878
104
XM_015290160.2
XP_015145646.1
8 - 9
1334
104
XM_015290161.2
XP_015145647.1
8 - 9
1305
104
XM_015290162.2
XP_015145648.1
8 - 9
1479
104
XM_015290163.2
XP_015145649.1
7 - 8
1217
104
XM_015290164.2
XP_015145650.1
7 - 8
1192
104
XM_015290165.2
XP_015145651.1
7 - 8
952
104
XM_015290166.2
XP_015145652.1
8 - 9
1101
104
XM_015290167.2
XP_015145653.1
7 - 8
744
104
XM_025152250.1
XP_025008018.1
8 - 9
1443
104
XM_025152252.1
XP_025008020.1
8 - 9
1518
104
XM_025152253.1
XP_025008021.1
8 - 9
1447
104
XM_025152254.1
XP_025008022.1
8 - 9
1309
104
XM_025152255.1
XP_025008023.1
8 - 9
1359
104
XM_025152256.1
XP_025008024.1
8 - 9
1069
104
XM_025152257.1
XP_025008025.1
8 - 9
1363
104
XM_025152258.1
XP_025008026.1
7 - 8
751
104
GenBank mRNA
AJ719461.1
-
7 - 8
1180
104
AJ726210.1
-
6 - 7
645
104
BU364244.1
-
3 - 4
165
104
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Detect Genomic DNA
Gallus gallus
Probe spans exons
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
ubiquitin-protein transferase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0