Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032398.4 | NP_001027570.3 | 7 - 8 | 2066 | 72 | |
XM_015296389.2 | XP_015151875.2 | 7 - 8 | 1377 | 72 | ||
XM_025142223.1 | XP_024997991.1 | 7 - 8 | 1444 | 72 | ||
GenBank mRNA | BU108973.1 | - | 2 - 3 | 377 | 72 | |
BX934090.2 | - | 5 - 6 | 1119 | 72 | ||
DR427690.1 | - | 4 - 5 | 663 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |