Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039601.1 | NP_001034690.1 | 26 - 27 | 3121 | 116 | |
XM_025152179.1 | XP_025007947.1 | 27 - 28 | 3175 | 116 | ||
XM_025152180.1 | XP_025007948.1 | 24 - 25 | 2988 | 116 | ||
GenBank mRNA | DQ150103.1 | - | 26 - 27 | 3121 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |