Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039601.1 | NP_001034690.1 | 28 - 29 | 3350 | 68 | |
XM_025152179.1 | XP_025007947.1 | 29 - 30 | 3404 | 68 | ||
XM_025152180.1 | XP_025007948.1 | 26 - 27 | 3217 | 68 | ||
GenBank mRNA | DQ150103.1 | - | 28 - 29 | 3350 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |