Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040464.1 | NP_001035554.1 | 2 - 3 | 144 | 68 | |
XM_015296359.2 | XP_015151845.1 | 2 - 3 | 4009 | 68 | ||
XM_015296360.2 | XP_015151846.1 | 3 - 4 | 491 | 68 | ||
GenBank mRNA | BX929984.1 | - | 2 - 3 | 260 | 68 | |
DQ465017.1 | - | 2 - 3 | 144 | 68 | ||
DQ465018.1 | - | 2 - 3 | 144 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |