Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044646.1 | NP_001038111.1 | 3 - 4 | 471 | 90 | |
XM_025148669.1 | XP_025004437.1 | 3 - 4 | 336 | 90 | ||
GenBank mRNA | AB253767.1 | - | 3 - 4 | 471 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |