Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044649.1 | NP_001038114.1 | 3 - 4 | 320 | 116 | |
XM_015285134.2 | XP_015140620.1 | 4 - 5 | 424 | 116 | ||
XM_015285135.2 | XP_015140621.1 | 4 - 5 | 457 | 116 | ||
XM_015285136.2 | XP_015140622.1 | 4 - 5 | 424 | 116 | ||
GenBank mRNA | AB253770.1 | - | 3 - 4 | 320 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |