Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044662.1 | NP_001038127.1 | 3 - 4 | 728 | 102 | |
XM_015290310.2 | XP_015145796.1 | 4 - 5 | 485 | 102 | ||
XM_015290311.2 | XP_015145797.1 | 4 - 5 | 608 | 102 | ||
XM_015290312.2 | XP_015145798.1 | 3 - 4 | 613 | 102 | ||
XM_015290313.2 | XP_015145799.1 | 4 - 5 | 485 | 102 | ||
XM_015290314.2 | XP_015145800.1 | 4 - 5 | 547 | 102 | ||
XM_015290315.2 | XP_015145801.1 | 4 - 5 | 608 | 102 | ||
GenBank mRNA | DQ640760.1 | - | 4 - 5 | 524 | 102 | |
DQ640761.1 | - | 3 - 4 | 728 | 102 | ||
DQ640762.1 | - | 4 - 5 | 524 | 102 | ||
DQ640763.1 | - | 3 - 4 | 618 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |