Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044665.1 | NP_001038130.1 | 1 - 2 | 129 | 77 | |
XM_015284842.2 | XP_015140328.1 | 2 - 3 | 8840 | 77 | ||
XM_015284843.2 | XP_015140329.1 | 2 - 3 | 352 | 77 | ||
GenBank mRNA | DQ129669.1 | - | 1 - 2 | 129 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |