Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044674.1 | NP_001038139.1 | 12 - 13 | 1356 | 62 | |
XM_015277359.1 | XP_015132845.1 | 12 - 13 | 1515 | 62 | ||
XM_015277360.2 | XP_015132846.1 | 12 - 13 | 1447 | 62 | ||
XM_015277361.2 | XP_015132847.1 | 12 - 13 | 1351 | 62 | ||
XM_015277362.2 | XP_015132848.1 | 12 - 13 | 1290 | 62 | ||
XM_015277363.2 | XP_015132849.1 | 12 - 13 | 1340 | 62 | ||
GenBank mRNA | AB161354.1 | - | 12 - 13 | 1356 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |