Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZPAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045837.1 | NP_001039302.1 | 17 - 18 | 2399 | 76 | |
XM_015284957.1 | XP_015140443.1 | 19 - 20 | 2568 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |