Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079494.1 | NP_001072962.1 | 10 - 11 | 1277 | 100 | |
XM_015291442.2 | XP_015146928.1 | 12 - 13 | 1267 | 100 | ||
XM_015291443.2 | XP_015146929.1 | 12 - 13 | 1337 | 100 | ||
XM_015291444.2 | XP_015146930.1 | 11 - 12 | 1276 | 100 | ||
XM_015291445.2 | XP_015146931.1 | 11 - 12 | 1284 | 100 | ||
XM_015291446.2 | XP_015146932.1 | 11 - 12 | 1348 | 100 | ||
XM_015291447.2 | XP_015146933.1 | 10 - 11 | 1262 | 100 | ||
XM_025153417.1 | XP_025009185.1 | 10 - 11 | 1292 | 100 | ||
GenBank mRNA | AJ719338.1 | - | 10 - 11 | 1277 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |