Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079495.2 | NP_001072963.1 | 7 - 8 | 948 | 83 | |
XM_015294248.2 | XP_015149734.1 | 7 - 8 | 948 | 83 | ||
XM_015294249.2 | XP_015149735.1 | 7 - 8 | 1019 | 83 | ||
XM_025155126.1 | XP_025010894.1 | 7 - 8 | 978 | 83 | ||
GenBank mRNA | AJ720447.1 | - | 7 - 8 | 950 | 83 | |
CR352740.1 | - | 7 - 8 | 876 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |