Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079739.1 | NP_001073207.1 | 1 - 2 | 382 | 104 | |
GenBank mRNA | AJ851659.1 | - | 1 - 2 | 382 | 104 | |
AM050146.1 | - | 1 - 2 | 87 | 104 | ||
EF554723.1 | - | 1 - 2 | 90 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |