Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079765.1 | NP_001073233.1 | 2 - 3 | 204 | 131 | |
GenBank mRNA | AJ720684.1 | - | 2 - 3 | 204 | 131 | |
CR389409.1 | - | 2 - 3 | 179 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |