Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079765.1 | NP_001073233.1 | 4 - 5 | 407 | 81 | |
GenBank mRNA | AJ720684.1 | - | 4 - 5 | 407 | 81 | |
CR389409.1 | - | 4 - 5 | 382 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |