Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080107.1 | NP_001073576.1 | 10 - 11 | 1259 | 104 | |
XM_015275342.2 | XP_015130828.1 | 10 - 11 | 1231 | 104 | ||
GenBank mRNA | EF118300.1 | - | 10 - 11 | 1259 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |