Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080870.1 | NP_001074339.1 | 384 | 116 | ||
XM_015298959.2 | XP_015154445.1 | 499 | 116 | |||
GenBank mRNA | AM076723.1 | - | 384 | 116 | ||
FR687371.1 | - | 1047 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus |