Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080891.1 | NP_001074360.1 | 5 - 6 | 743 | 94 | |
GenBank mRNA | AJ720597.1 | - | 5 - 6 | 743 | 94 | |
CR406324.1 | - | 3 - 4 | 473 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |