Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081504.1 | NP_001074973.1 | 2 - 3 | 305 | 64 | |
XM_015299821.2 | XP_015155307.1 | 3 - 4 | 498 | 64 | ||
XM_015299822.2 | XP_015155308.1 | 3 - 4 | 589 | 64 | ||
XM_025144283.1 | XP_025000051.1 | 2 - 3 | 10053 | 64 | ||
GenBank mRNA | AB257073.1 | - | 2 - 3 | 305 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |