Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083359.1 | NP_001076828.1 | 8 - 9 | 1504 | 74 | |
XM_015275308.2 | XP_015130794.1 | 9 - 10 | 2436 | 74 | ||
GenBank mRNA | EF141825.1 | - | 8 - 9 | 1504 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |