Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083360.1 | NP_001076829.1 | 2 - 3 | 156 | 109 | |
GenBank mRNA | DQ360501.1 | - | 2 - 3 | 156 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |