Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097537.1 | NP_001091006.1 | 2 - 3 | 258 | 92 | |
XM_015294250.2 | XP_015149736.1 | 3 - 4 | 550 | 92 | ||
XM_015294251.2 | XP_015149737.1 | 3 - 4 | 566 | 92 | ||
XM_015294252.2 | XP_015149738.1 | 3 - 4 | 514 | 92 | ||
GenBank mRNA | AM492777.1 | - | 2 - 3 | 258 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |