Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THOC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098238.3 | NP_001091708.3 | 5 - 6 | 441 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |