Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098608.1 | NP_001092078.1 | 4 - 5 | 430 | 109 | |
XM_025153365.1 | XP_025009133.1 | 4 - 5 | 531 | 109 | ||
XM_025153367.1 | XP_025009135.1 | 4 - 5 | 562 | 109 | ||
GenBank mRNA | CR389564.1 | - | 4 - 5 | 456 | 109 | |
EF551041.1 | - | 4 - 5 | 430 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |