Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271901.1 | NP_001258830.1 | 7 - 8 | 1151 | 67 | |
NM_001271902.1 | NP_001258831.1 | 7 - 8 | 1048 | 67 | ||
XM_015289786.2 | XP_015145272.1 | 7 - 8 | 1366 | 67 | ||
XM_025152142.1 | XP_025007910.1 | 8 - 9 | 1298 | 67 | ||
GenBank mRNA | AF022818.1 | - | 7 - 8 | 1052 | 67 | |
AM262745.1 | - | 7 - 8 | 1114 | 67 | ||
AM262746.1 | - | 7 - 8 | 1151 | 67 | ||
AM262747.1 | - | 5 - 6 | 950 | 67 | ||
U60763.1 | - | 3 - 4 | 516 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |