Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098853.1 | NP_001092323.1 | 4 - 5 | 646 | 89 | |
XM_015287569.2 | XP_015143055.1 | 5 - 6 | 3767 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |