Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM7.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099352.1 | NP_001092822.1 | 3 - 4 | 219 | 85 | |
XM_015294955.1 | XP_015150441.1 | 3 - 4 | 996 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |