Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101034.1 | NP_001094504.1 | 2 - 3 | 656 | 103 | |
GenBank mRNA | BX930481.1 | - | 1 - 2 | 113 | 103 | |
EF624351.1 | - | 2 - 3 | 656 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |