Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105315.1 | NP_001098785.1 | 3 - 4 | 265 | 113 | |
GenBank mRNA | DQ870922.1 | - | 3 - 4 | 265 | 113 | |
DQ870923.1 | - | 2 - 3 | 101 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |