Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPNVA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105318.1 | NP_001098788.1 | 3 - 4 | 541 | 108 | |
NM_001310089.1 | NP_001297018.1 | 2 - 3 | 572 | 108 | ||
NM_001310090.1 | NP_001297019.1 | 2 - 3 | 572 | 108 | ||
XM_015288991.2 | XP_015144477.1 | 5 - 6 | 1454 | 108 | ||
XM_015288992.1 | XP_015144478.1 | 2 - 3 | 741 | 108 | ||
GenBank mRNA | AB295600.1 | - | 3 - 4 | 541 | 108 | |
EF055883.2 | - | 2 - 3 | 572 | 108 | ||
GQ280390.1 | - | 2 - 3 | 572 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Gallus gallus Probe spans exons |