Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109679.1 | NP_001103149.1 | 2 - 3 | 727 | 87 | |
GenBank mRNA | CR406467.1 | - | 2 - 3 | 169 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |