Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109692.1 | NP_001103162.1 | 6 - 7 | 594 | 116 | |
XM_015282328.2 | XP_015137814.1 | 5 - 6 | 464 | 116 | ||
GenBank mRNA | CR386184.1 | - | 6 - 7 | 565 | 116 | |
CR387537.1 | - | 6 - 7 | 524 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |