Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109787.1 | NP_001103257.1 | 12 - 13 | 1611 | 98 | |
GenBank mRNA | X67786.1 | - | 1611 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |