Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110060.1 | NP_001103530.1 | 2 - 3 | 244 | 104 | |
XM_015284828.2 | XP_015140314.1 | 3 - 4 | 436 | 104 | ||
XM_015284829.2 | XP_015140315.1 | 3 - 4 | 452 | 104 | ||
GenBank mRNA | BX929572.2 | - | 3 - 4 | 389 | 104 | |
BX931809.1 | - | 3 - 4 | 406 | 104 | ||
CR406263.1 | - | 3 - 4 | 394 | 104 | ||
EU176872.1 | - | 2 - 3 | 244 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |