Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190165.4 | NP_001177094.1 | 6 - 7 | 701 | 137 | |
NR_073182.3 | 6 - 7 | 690 | 137 | |||
XM_015289639.2 | XP_015145125.1 | 7 - 8 | 4472 | 137 | ||
GenBank mRNA | DQ185931.1 | - | 6 - 7 | 701 | 137 | |
DR429295.1 | - | 6 - 7 | 697 | 137 | ||
S78477.1 | - | 6 - 7 | 699 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |