Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205174.2 | NP_990505.1 | 4 - 5 | 1681 | 110 | |
GenBank mRNA | S53899.1 | - | 4 - 5 | 1681 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |