Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205308.2 | NP_990639.1 | 3 - 4 | 352 | 83 | |
GenBank mRNA | X65459.1 | - | 352 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |