Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113291.1 | NP_001106762.1 | 5 - 6 | 806 | 153 | |
XM_015286619.2 | XP_015142105.2 | 5 - 6 | 602 | 153 | ||
GenBank mRNA | CR386788.1 | - | 4 - 5 | 438 | 153 | |
EU240627.2 | - | 5 - 6 | 806 | 153 | ||
EU852334.1 | - | 5 - 6 | 772 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |