Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113291.1 | NP_001106762.1 | 6 - 7 | 981 | 86 | |
XM_015286619.2 | XP_015142105.2 | 6 - 7 | 777 | 86 | ||
GenBank mRNA | EU240627.2 | - | 6 - 7 | 981 | 86 | |
EU419874.1 | - | 6 - 7 | 930 | 86 | ||
EU852334.1 | - | 6 - 7 | 947 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |