Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114852.1 | NP_001108324.1 | 3 - 4 | 203 | 122 | |
XM_015276165.2 | XP_015131651.1 | 3 - 4 | 1425 | 122 | ||
XM_015276166.2 | XP_015131652.1 | 3 - 4 | 366 | 122 | ||
GenBank mRNA | CR385387.1 | - | 3 - 4 | 361 | 122 | |
EU118175.1 | - | 3 - 4 | 203 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |